missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL2RG Recombinant Rat Monoclonal Antibody (3E12)
GREENER_CHOICE

Catalog No. MA551816
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551816 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551816 Supplier Invitrogen™ Supplier No. MA551816
Only null left

Rat Recombinant Monoclonal Antibody

This conformation-dependent antibody binds to the murine common gamma-chain, shared by the IL-2, IL-4, IL-7, IL-9, IL-15, and IL-21 receptor complexes. It recognises an epitope located in a membrane-proximal portion of the common gamma-chain, near Arg-197 of domain 2. This antibody does not cross-react with COS7 cells transfected with rat common gamma-chain cDNA.

IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.

Specifications

Antigen IL2RG
Applications Flow Cytometry, Inhibition Assays, Immunoprecipitation
Classification Recombinant Monoclonal
Clone 3E12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene IL2RG
Gene Accession No. P34902
Gene Alias [g]c; Ab2-183; CD132; CD132 antigen; CIDX; common cytokine receptor gamma chain; cytokine receptor common subunit gamma; gamma(c); gammaC; gc; IL-2 receptor subunit gamma; IL2R gamma; IL-2R subunit gamma; IL2RG; IL-2RG; IMD4; interleukin 2 receptor subunit gamma; interleukin 2 receptor, gamma; interleukin 2 receptor, gamma (severe combined immunodeficiency); interleukin 2 receptor, gamma chain; Interleukin-2 receptor subunit gamma; p64; SCIDX; SCIDX1
Gene Symbols IL2RG
Host Species Rat
Immunogen This antibody was raised by immunising Lewis rats with transfected rat myeloma cell line YB2/0, expressing murine cytoplasmic-tailless IL-2R common gamma-chain (YB2/0-myt).
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16186
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less