missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL23R Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579490
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579490 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579490 Supplier Invitrogen™ Supplier No. PA579490
Only null left

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: mouse brain tissue, mouse liver tissue, mouse kidney tissue.

IL-23R is a 629 aa, novel subunit of the receptor for cytokine IL23A/IL23 which pairs with the receptor molecule IL12RB1/IL12R beta 1and thus take part in IL23A signaling. Signal transduction analysis confirms that this protein associates constitutively with Janus kinase 2 (JAK2), and also binds to transcription activator STAT3 in a ligand-dependent manner. Alternative splicing results in six spliced isoforms of IL-23R. Reports suggest that IL-23R plays an important role in psoriasis pathogenesis and is also a potential therapeutic target in inflammatory bowel disease.

Specifications

Antigen IL23R
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene Il23r
Gene Accession No. Q8K4B4
Gene Alias IBD17; IL 23R; IL-23 receptor; IL23R; IL-23R; interleukin 23 receptor; interleukin 23 receptor-like; interleukin-23 receptor
Gene Symbols Il23r
Host Species Rabbit
Immunogen E. coli-derived mouse IL23 Receptor recombinant protein (Position: I25-D233).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209590, 689932
Target Species Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less