missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-6 Monoclonal Antibody (MP5-20F3), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
GREENER_CHOICE

Rat Monoclonal Antibody

Supplier:  Invitrogen™ 46706182

 View more versions of this product

Catalog No. 501123888


Only null left

Description

Description

Description: The MP5-20F3 antibody reacts with mouse interleukin-6 (IL-6). The MP5-20F3 antibody is a neutralizing antibody. Mouse IL-6 (also known as B Cell Stimulating Factor, BCSF, and BSF-2) is a 21-28 kDa factor produced by a variety of cell types, including monocytes, fibroblasts and endothelial cells. Upon stimulation, macrophages, T, B, mast, and glial cells, eosinophils, keratinocytes and granulocytes also secrete IL-6. This pleiotrophic cytokine influences the antigen-specific immune response, inflammatory response, and the development of neurons, and is a major mediator of the acute phase reaction. Applications Reported: This MP5-20F3 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This MP5-20F3 antibody has been tested by intracellular staining and flow cytometric analysis of stimulated mouse thioglycolate-elicited peritoneal exudate cells using the Intracellular Fixation & Permeabilization Buffer Set (cat. 88-8824) and protocol. Please refer to Best Protocols: Protocol A: Two step protocol for (cytoplasmic) intracellular proteins located under the Resources Tab online. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.

IL-6 is a gene that encodes a cytokine that plays a role in inflammation and the maturation of B cells. The protein produced by this gene is an endogenous pyrogen that can induce fever in people with autoimmune diseases or infections. The gene is primarily expressed at sites of acute and chronic inflammation, where it is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of this gene is implicated in a wide variety of inflammation-associated disease states, including suspectibility to diabetes mellitus and systemic juvenile rheumatoid arthritis. Elevated levels of the encoded protein have been found in virus infections, including COVID-19 (disease caused by SARS-CoV-2). Diseases associated with IL6 include Kaposi Sarcoma and Rheumatoid Arthritis, Systemic Juvenile.
Specifications

Chemical Identifiers

IL-6
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6
Il6
Primary
4°C, store in dark, DO NOT FREEZE!
Il6
MP5-20F3
Flow Cytometry
PerCP-eFluor 710
Rat
Mouse
P08505
16193
IgG1 κ
Affinity chromatography
Antibody

Specifications

IL-6
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P08505
Il6
Affinity chromatography
RUO
16193
4°C, store in dark, DO NOT FREEZE!
Liquid
Flow Cytometry
MP5-20F3
PerCP-eFluor 710
Il6
B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; CHIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; hybridoma growth factor; Ifnb2; IFN-beta-2; Il6; IL-6; ILg6; ILN; interferon beta-2; Interleukin; interleukin 6; Interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6
Rat
100 μg
Primary
Mouse
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only.