missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ IL-6 ELISPOT Antibody Pair, Mouse

Catalog No. ELI008M
Change view
Click to view available options
Quantity:
96-well Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ELI008M 96-well Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ELI008M Supplier MilliporeSigma™ Supplier No. ELI008M

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Protein Interaction Antibody Pairs

Includes:

  • Mouse IL-6 Capture Antibody, clone MP5-20F3. Quantity: 500 μg.
  • Mouse IL-6 Detection Antibody, clone MP5-32C11. Quantity: 100 μg.
  • Streptavidin Alkaline Phosphatase. Quantity: 55 μL.

Specifications

Classification Monoclonal
Concentration 1.0mg/mL (Capture antibody), 0.5mg/mL (Biotinylated detection antibody)
Host Species Mouse
Quantity 96-well Plates
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for up to six months
Product Type MP5-20F3 (Capture), MP5-32C11 (Detection)
Form Purified