missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IL-5 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. 500P55BT25
Encompass
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P55BT25 25 μg
500P55BT50 50 μg
502718472 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 500P55BT25 Supplier PeproTech Supplier No. 500P55BT25UG
Only null left

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MEIPMSTVVK ETLTQLSAHR ALLTSNETMR LPVPTHKNHQ LCIGEIFQGL DILKNQTVRG GTVEMLFQNL SLIKKYIDRQ KEKCGEERRR TRQFLDYLQE FLGVMSTEWA MEG. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-5. Anti-Murine IL-5-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Murine IL-5 by sandwich ELISA (using 100 mL/well) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti- Murine IL-5 (500-P55) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Murine IL-5. Western Blot: To detect Murine IL-5 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 mg/mL. When used in conjunction with compatible development reagents, the detection limit for Recombinant Murine IL-5 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P55BT-1MG will be provided as 2 x 500 μg

IL-5 is a cytokine that acts as a growth and differentiation factor for both B cells and eosinophils. IL-5 cytokine is a main regulator of eosinopoiesis, eosinophil maturation and activation. The elevated production of this cytokine is reported to be related to asthma or hypereosinophilic syndromes. The receptor of IL-5 cytokine is a heterodimer, whose beta subunit is shared with the receptors for interleukine 3 (IL3) and colony stimulating factor 2 (CSF2/GM-CSF). IL-5, together with those for interleukin 4 (IL4), interleukin 13 (IL13), and CSF2, form a cytokine gene cluster on chromosome 5. IL-5, IL4, and IL13 are found to be regulated coordinately by long-range regulatory elements spread over 120 kilobases on chromosome 5q31.

Specifications

Antigen IL-5
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene Il5
Gene Accession No. P04401
Gene Alias B-cell differentiation factor I; B-cell growth factor II; BCGF-II; colony-stimulating factor, eosinophil; cytotoxic T-lymphocyte inducer; EDF; Eosinophil differentiation factor; IL5; IL-5; ILN; Interleukin; Interleukin 5; Interleukin 5 (colony-stimulating factor eosinophil); Interleukin 5 (colony-stimulating factor, eosinophil); Interleukin5; interleukin-5; T-cell replacing factor; TRF
Gene Symbols Il5
Host Species Rabbit
Immunogen E.coli-derived Recombinant Murine IL-5.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16191
Target Species Mouse
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less