Learn More
Invitrogen™ IL-4 Monoclonal Antibody (OX81), eFluor™ 660, eBioscience™

Description
Description: This OX81 monoclonal antibody reacts with rat interleukin (IL)-4. Also known as BCDF, BCGF-1 and BSF-1, IL-4 is secreted by activated Th2 and natural killer T cells. This cytokine is also produced by Th1 and mast cells albeit to a lesser extent. IL-4 is species-specific and promotes proliferation and differentiation of B cells. IL-4, in conjunction with other signals such as CD40, induces Ig isotype switching of B cells and the upregulation of MHC class II and CD23. The OX81 antibody has been reported to exhibit neutralizing activity. Applications Reported: This OX81 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This OX81 antibody has been tested by intracellular flow cytometric analysis of activated rat splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manuf...
Specifications
Specifications
| Antigen | IL-4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OX81 |
| Concentration | 0.2 mg/mL |
| Conjugate | eFluor 660 |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Il4 |
| Gene Accession No. | P20096 |
| Gene Alias | B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra |
| Show More |