Learn More
Invitrogen™ IL-4 Monoclonal Antibody (11B11), Functional Grade, eBioscience™, Invitrogen™

Description
Description: The 11B11 antibody reacts with mouse interleukin-4 (IL-4), a 14 kD cytokine. The 11B11 antibody is a neutralizing antibody. Mouse IL-4, also known as BCDF, BCGF-1 and BSF-1, is secreted by activated Th2 and natural killer T cells, and to a lesser extent by Th1 and mast cells. IL-4 is species-specific and promotes proliferation and differentiation of B cells. IL-4, in conjunction with other signals such as CD40, induces Ig isotype switching of B cells and the upregulation of surface MHC class II and CD23. Applications Reported: This 11B11 antibody has been reported for use in cytokine neutralization, ELISPOT, and ELISA cap. Applications Tested: The Functional Grade Purified 11B11 antibody has been tested by LAL assay to verify low endotoxin levels, by ELISA, and in bioassay for neutralization of mouse IL-4 bioactivity. The 11B11 antibody at 0.125 μg/mL has been found to inhibit by 50% the biological effects of 1.0 ng/mL mouse IL-4 in a CTLL-2 cell proliferation assay. Please refer to Best Protocols for more information. The 11B11 antibody has been tested as the capture antibody in a sandwich ELISA for analysis of mouse interleukin-4 (IL-4) in combination with the biotinylated BVD6-24G2 (cat. 13-7042) antibody for detection and recombit mouse IL-4 (cat. 14-8041) as the standard. A suitable range of concentrations of this antibody for ELISA capture is 1-4 μg/mL.
Specifications
Specifications
| Antigen | IL-4 |
| Applications | Functional Assay, Immunostaining |
| Classification | Monoclonal |
| Clone | 11B11 |
| Concentration | 1 mg/mL |
| Conjugate | Functional Grade |
| Formulation | PBS with no preservative; pH 7.2 |
| Gene | Il4 |
| Gene Accession No. | P07750 |
| Gene Alias | B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4delta2; interleukin 4 variant IL-4delta3; interleukin 4 variant IL-4int2A; interleukin 4 variant IL-4int3A; interleukin 4 variant IL-4int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra |
| Show More |
For Research Use Only.