missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-37 Monoclonal Antibody (K92047_1H12), Invitrogen™

Catalog No. MA556096
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556096 200 μL
MA555716 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556096 Supplier Invitrogen™ Supplier No. MA556096
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: KVKNLNPKKF SIHDQDHKVL VLDSGNLIAV PDKNYIRPEI FFALASSLSS ASAEKGSPIL LGVSKGEFCL YCDKDKGQSH PSLQLKKEKL

The protein encoded by this gene is a member of the interleukin 1 cytokine family. This cytokine can bind to, and may be a ligand for interleukin 18 receptor (IL18R1/IL-1Rrp). This cytokine also binds to interleukin 18 binding protein (IL18BP), an inhibitory binding protein of interleukin 18 (IL18), and subsequently forms a complex with IL18 receptor beta subunit, and through which it inhibits the activity of IL18. This gene along with eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2. Five alternatively spliced transcript variants encoding distinct isoforms have been reported.

Specifications

Antigen IL-37
Applications Western Blot
Classification Monoclonal
Clone K92047_1H12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene IL37
Gene Accession No. Q9NZH6
Gene Alias FIL1; FIL1 zeta; FIL1(ZETA); FIL1Z; IL-1 zeta; IL1F7; IL-1F7; IL1F7 (canonical product IL-1F7b); IL-1F7b (IL-1H4, IL-1H, IL-1RP1); IL-1H; IL1H4; IL-1H4; IL1RP1; IL-1RP1; IL-1X; IL-1X protein; IL37; IL-37; ILN; Interleukin; interleukin 1 family member 7; interleukin 1, zeta; interleukin 37; Interleukin-1 family member 7; Interleukin-1 homolog 4; interleukin-1 superfamily z; Interleukin-1 zeta; Interleukin-1-related protein; interleukin-23; Interleukin37; Interleukin-37
Gene Symbols IL37
Host Species Mouse
Immunogen Recombinant human IL37 (immunogen range is 51-140).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27178
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less