missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL-33R (ST2) Monoclonal Antibody (hIL33Rcap), Brilliant Violet™ 421, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$624.38 - $1322.02
Specifications
| Antigen | IL-33R (ST2) |
|---|---|
| Clone | hIL33Rcap |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
40-493-3842
|
Invitrogen™
404933842 |
100 Tests |
Each for $1,322.02
|
|
|||||
|
40-493-3841
|
Invitrogen™
404933841 |
25 Tests |
Each for $624.38
|
|
|||||
Description
IL-33R (ST2) Monoclonal antibody specifically detects IL-33R (ST2) in Human samples. It is validated for Flow CytometrySpecifications
| IL-33R (ST2) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| DER4; Fit1; Fit-1; Fos-responsive gene 1 protein; growth stimulation-expressed; homolog of mouse growth stimulation-expressed; IL-1 R4; IL1RL1; IL-1RL1; IL33R; IL-33R; interleukin 1 receptor like 1; interleukin 1 receptor-like 1; interleukin 1 receptor-related protein; Interleukin1 receptor like 1; interleukin-1 receptor-like 1; Interleukin-33 receptor alpha chain; Ly84; lymphocyte antigen 84; Protein ST2; Protein T1; sIL 33R; soluble IL 33 Receptor; soluble IL 33R; St2; ST2L; St2-rs1; ST2V; Ste2; T1; T1/ST2 | |
| Extracellular domain of human IL-33R protein. | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| IL1RL1 |
| hIL33Rcap | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| Mouse | |
| Human | |
| Q01638 | |
| 9173 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |