missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-3 ProQuantum Immunoassay Kit

Description
Interleukin 3 (IL-3), also called Mast Cell Growth Factor (MCGF), Multi-Colony Stimulating Factor (multi-CSF), or Eosinophil-CSF (E-CSF), is produced by activated T cells, mast cells and eosinophils. IL-3 is a hematopoietic growth factor that stimulates colony formation of erythroid, megakaryocyte, neutrophil, eosinophil, basophil, mast cell and monocytic lineages. Most of these functions are enhanced or dependent on co-stimulation with other cytokines. Human and mouse IL-3 share only 29% sequence identity at the amino acid level. The bioactivity of IL-3 is highly species specific. For example, human IL-3 has no activity on mouse cells. IL-3 is a potent growth promoting cytokine. IL-3 is capable of supporting the proliferation of a broad range of hematopoietic cell types, and is involved in a variety of cell activities such as cell growth, differentiation and apoptosis. IL-3 has been shown to also possess neurotrophic activity, and it may be associated with neurologic disorders.
Specifications
Specifications
| Accession Number | P08700 |
| Assay Range | 0.0128 to 5000 pg/mL |
| Assay Sensitivity | <0.01 pg/mL |
| Conjugate | FAM™ |
| Detection Method | Taqman™ Real-time PCR |
| Immunoassay Kit Format | Proximity-based Immunoassay |
| Product Type | ProQuantum |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Application) | Protein Quantitation |
| Show More |