missing translation for 'onlineSavingsMsg'
Learn More

IL-26 Polyclonal Antibody

IL-26 Polyclonal Antibody

Supplier:  Thermo Fisher Scientific PA526482

Catalog No. PIPA526482


Only null left
Explore available promotions

Description

Description

IL-26 Polyclonal Antibody for Western Blot

Specifications

Specifications

IL-26
Polyclonal
Unconjugated
IL26
AK155; il 26; IL26; IL-26; ILN; Interleukin; interleukin 26; interleukin-26; Protein AK155
Rabbit
Antigen Affinity Chromatography, Protein A
RUO
55801
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.25 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q9NPH9
IL26
KLH conjugated synthetic peptide between 88-115 amino acids from the central region of human IL26.
400 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions