missing translation for 'onlineSavingsMsg'
Learn More

PeproTech IL-25 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. 500P89BT25
Encompass
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P89BT25 25 μg
500P89BT50 50 μg
502718436 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 500P89BT25 Supplier PeproTech Supplier No. 500P89BT25UG
Only null left

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MYSHWPSCCP SKGQDTSEEL LRWSTVPVPP LEPARPNRHP ESCRASEDGP LNSRAISPWR YELDRDLNRL PQDLYHARCL CPHCVSLQTG SHMDPRGNSE LLYHNQTVFY RRPCHGEKGT HKGYCLERRL YRVSLACVCV RPRVMG. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human IL-17E. Anti-Human IL-17E-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human IL-17E by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human IL-17E (500-P89Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human IL-17E. Western Blot: To detect Human IL-17E by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL.andnbsp; When used in conjunction with compatible secondary reagents the detection limit for Recombinant Human IL-17E is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P89BT-1MG will be provided as 2 x 500 μg

Interleukin 25 (IL-25) is a member of the IL-17 family and is identified as a proinflammatory cytokine which is produced by Th17 effector cells thus enabling the Th17 subset to initiate or maintain the autoimmune process in diseases such as Rheumatoid Arthritis. IL-25 also induces activation of NF-kappa-B and stimulates production of the proinflammatory chemokine IL-8. IL-25 itself is activated through IL-23, IL-5, and TGF-beta.

Specifications

Antigen IL-25
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene IL25
Gene Accession No. Q9H293
Gene Alias D17Wsu104e; il 17e; Il17e; IL-17E; Il25; IL-25; ILN; Interleukin; interleukin 17E; interleukin 25; interleukin-17E; Interleukin-25; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; RGD1561632; UNQ3120/PRO10272; UPF0556 protein C19orf10 homolog
Gene Symbols IL25
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human IL-17E.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64806
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less