missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse IL-2 ProQuantum Immunoassay Kit

ELISA
$948.00 - $4,280.00
Specifications
| Accession Number | P04351 |
|---|---|
| Assay Range | 0.064 to 5000 pg/mL |
| Assay Sensitivity | <0.05 pg/mL |
| Conjugate | FAM™ |
| Detection Method | Taqman™ Real-time PCR |
Description
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Specifications
| P04351 | |
| <0.05 pg/mL | |
| Taqman™ Real-time PCR | |
| ProQuantum | |
| Mouse | |
| qPCR Instruments | |
| 1 hr. | |
| FAM™ | |
| Plasma 2-5 μL, Serum 2-5 μL, Supernatant 2-5 μL | |
| IL-2 | |
| 2 hrs |
| 0.064 to 5000 pg/mL | |
| FAM™ | |
| Proximity-based Immunoassay | |
| Plasma, Serum, Supernatant | |
| Protein Quantitation | |
| 16183 | |
| Assay Dilution Buffer, Antibody-Conjugate A, Antibody-Conjugate B, Antibody-Conjugate Dilution Buffer, Standard, Master Mix, Ligase | |
| RUO | |
| Frozen at -20°C | |
| 30 mins |