Learn More
Invitrogen™ IL-2 Monoclonal Antibody (JES6-5H4), FITC, eBioscience™, Invitrogen™
Description
Description: The JES6-5H4 antibody reacts with mouse interleukin-2 (IL-2), a 17 kDa T cell growth factor and a major immunoregulatory cytokine. Applications Reported: JES6-5H4 has been reported for use in intracellular staining and flow cytometric analysis. Applications Tested: The JES6-5H4 is offered in 2 formats: - μg size: has been tested by intracellular staining and flow cytometric analysis and can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. - test size: has been pre-titrated and tested by intracellular staining and flow cytometric analysis. Refer to catalog number suffix on the vial for amount to use per test: -41 is 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | IL-2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | JES6-5H4 |
| Concentration | 0.5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | IL2 |
| Gene Accession No. | P04351 |
| Gene Alias | aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF |
| Show More |
For Research Use Only.