Learn More
Invitrogen™ Equine IL-1RA ELISA Kit
ELISA
$942.00
Specifications
| Accession Number | O18999 |
|---|---|
| Assay Range | 12.29 to 3000 pg/mL |
| Assay Sensitivity | 10 pg/mL |
| Calibration Range | 10 pg/mL to 3000 pg/mL |
| Assay | Solid-phase sandwich ELISA |
Description
The Equine Interleukin-1 Receptor Antagonist (Eq IL-1RA) ELISA quantitates Eq IL-1RA in equine serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Eq IL-1RA. Principle of the method The Equine IL-1RA solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is a cytokine receptor that belongs to the interleukin 1 receptor family. This protein is a receptor for interleukin alpha (IL1A), interleukin beta (IL1B), and interleukin 1 receptor, type I(IL1R1/IL1RA). It is an important mediator involved in many cytokine induced immune and inflammatory responses. This gene along with interleukin 1 receptor, type II (IL1R2), interleukin 1 receptor-like 2 (IL1RL2), and interleukin 1 receptor-like 1 (IL1RL1) form a cytokine receptor gene cluster in a region mapped to chromosome 2q12.Specifications
| O18999 | |
| 10 pg/mL | |
| Solid-phase sandwich ELISA | |
| ELISA | |
| Plasma, Serum, Supernatant | |
| 96 Assays | |
| Colorimetric Microplate Reader | |
| IL-1 RA | |
| <10 | |
| HRP | |
| Extracellular Matrix & Cell Adhesion | |
| 2°C to 8°C | |
| IL1RA | |
| 4 hrs 45 mins |
| 12.29 to 3000 pg/mL | |
| 10 pg/mL to 3000 pg/mL | |
| HRP | |
| Cell Structure & Organelle Proteins | |
| The sandwich ELISA antibody pair detects Equine IL-1 RA. | |
| ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
| 100034236 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| Plasma 33 μL, Serum 33 μL, Supernatant 100 μL | |
| IL-1 RA | |
| 1 hrs 20 mins |