missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-18RB/IL-18RAP ELISA Kit

ELISA

$890.00

Specifications

Accession Number O95256
Assay Range 12.29 to 3000 pg/mL
Assay Sensitivity 12 pg/mL
Calibration Range 12 pg/mL to 3000 pg/mL
Assay Solid-phase sandwich ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EHIL18RAP
Encompass
View Documents Promotion Details
Invitrogen™
EHIL18RAP
96 Tests
Pack for $890.00
Only null left
 
Description

Description

The Human Interleukin 18 Receptor Beta (Hu IL-18RB) (IL-18RAP) ELISA quantitates Hu IL-18RB in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-18RB. Principle of the method The Human IL-18RB solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The protein encoded by this gene is an accessory subunit of the heterodimeric receptor for IL18. This protein enhances the IL18 binding activity of IL18R1 (IL1RRP), a ligand binding subunit of IL18 receptor. The coexpression of IL18R1 and this protein is required for the activation of NF-kappaB and MAPK8 (JNK) in response to IL18.
Specifications

Specifications

O95256
12 pg/mL
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
Human
Colorimetric Microplate Reader
ACPL,CD218b,CDw218b,IL-18 R-beta,IL-18 RAcP,IL-18 Rbeta,IL-1 R-7,IL-1R7,IL-1 RAcPL,IL18RB
<10
HRP
Inflammation
2°C to 8°C
IL18RAP
4 hrs 45 mins
12.29 to 3000 pg/mL
12 pg/mL to 3000 pg/mL
HRP
Other Proteins
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, ENA- 78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL- 11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF- 7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, SDF-1 alpha, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
8807
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
ACPL, CD218b, CDw218b, IL-18 R-beta, IL-18 RAcP, IL-18 Rbeta, IL-1 R-7, IL-1R7, IL-1 RAcPL, IL18RB
1 hrs 20 mins
Videos
SDS
Documents

Documents