Learn More
Invitrogen™ Mouse IL-17AF (heterodimer) Uncoated ELISA Kit
ELISA
Supplier: Invitrogen™ 88871177
Description
The Mouse Interleukin 17AF (heterodimer) (IL-17AF) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IL-17AF(heterodimer). Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
IL-17A and IL-17F are well-characterized homodimeric cytokines secreted by T helper 17 (Th17) cells, gamma-delta T cells and several subsets of innate lymphoid cells. Somewhat less appreciated, IL-17A and IL-17F subunits can also form the heterodimer, IL-17AF. Together, these three dimers signal through the IL-17RA/IL-17RC receptor complex to mediate immune responses at mucosal interfaces and are found at lesion sites in inflammatory bowel disease, asthma, atopic dermatitis and rheumatoid arthritis.Specifications
| Q62386,Q7TNI7 | |
| 16 pg/ml | |
| ELISA | |
| Mouse | |
| 16171, 257630 | |
| Capture Antibody: Pre-titrated, Purified antibody, Detection Antibody: Pre-titrated, Purified antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, Biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, Sample Diluent A: 12 mL of a 1X solution per plate, Assay Diluent: 5X concentrated, Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution, Detection enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution | |
| 20 x 96 Tests | |
| Serum 50 μL, Supernatant 50 μL | |
| IL-17 AF | |
| 24 hrs 30 mins |
| 16 to 2,000 pg/mL | |
| HRP | |
| Serum, Supernatant | |
| Colorimetric Microplate Reader | |
| GCP-1,GCP1,IL8,LECT,LUCT,LYNAP,MDNCF,MONAP,NAF,NAP-1,NAP1 | |
| HRP | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 45 mins |