Learn More
Invitrogen™ Human IL-17A (homodimer) Uncoated ELISA Kit
ELISA
$1,066.00 - $1,468.00
Specifications
| Accession Number | Q16552 |
|---|---|
| Assay Range | 4 to 500 pg/mL |
| Assay Sensitivity | 4 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
Description
The Human Interleukin 17A (IL-17A) Uncoated ELISA Kit contains pre-matched antibody pairs, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human IL-17A. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).Specifications
| Q16552 | |
| 4 pg/mL | |
| ELISA | |
| Human | |
| Colorimetric Microplate Reader | |
| CANDF6,IL-17 F,ML-1,ML1 | |
| HRP | |
| Serum 100 μL, Supernatant 100 μL | |
| Interleukin-17 A, IL-17 AA | |
| 1 hrs 45 mins |
| 4 to 500 pg/mL | |
| HRP | |
| Serum, Supernatant | |
| ELISA | |
| 3605 | |
| Capture Antibody: Pre-titrated, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86), Purified antibody, Biotin-conjugated antibody, Biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, Assay Diluent: 5X concentrated, Assay Diluent: 5X concentrated, Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards | |
| RUO | |
| 2°C to 8°C | |
| IL-17 A | |
| 24 hrs 30 mins |