missing translation for 'onlineSavingsMsg'
Learn More

IL-16 Monoclonal Antibody (K1E006_5C3), Invitrogen™

Mouse Monoclonal Antibody

$1740.58 - $2926.56

Specifications

Antigen IL-16
Clone K1E006_5C3
Concentration 1 mg/mL
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MA555936
View Documents
Invitrogen™
MA555936
50 μL
Each for $1,740.58
Only null left
 
MA556316
View Documents
Invitrogen™
MA556316
200 μL
Each for $2,926.56
Only null left
 
Description

Description

Sequence of this protein is as follows: DSAASASAAS DVSVESTAEA TVCTVTLEKM SAGLGFSLEG GKGSLHGDKP LTINRIFKGA ASEQSETVQP GDEILQLGGT AMQGLTRFEA WNIIKALPDG PVTIVIRRKS LQSKETTAAG

IL-16 Monoclonal antibody specifically detects IL-16 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

IL-16
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
FLJ16806; FLJ42735; FLJ44234; H-IL-16; il 16; Il16; IL-16; ILN; Interleukin; interleukin 16; Interleukin16; Interleukin-16; LCF; Lymphocyte chemoattractant factor; M-IL-16; mKIAA4048; neuronal interleukin 16; NIL16; PRIL16; prIL-16; prointerleukin 16; Pro-interleukin-16
Recombinant human IL16 (immunogen range is 1211-1330).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IL16
K1E006_5C3
ELISA, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human
Q14005
3603
IgG2a
Protein A
Antibody
Videos
SDS
Documents

Documents