Learn More
Invitrogen™ Human IL-10 ELISA Kit
ELISA
Supplier: Invitrogen™ EHIL105
Description
The Human Interleukin 10 (Hu IL-10) ELISA quantitates Hu IL-10 in human serum, plasma, urine, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-10. Principle of the method The Human IL-10 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.Specifications
| P22301 | |
| <3 pg/mL | |
| HRP | |
| ELISA | |
| No significant cross-reaction with human IL-1 A, IL-2, IL-3, IL-4, IL-6, IL-7, IL-8, TNFa, TNFÃ or IFNg. Approx. 2% cross-reactivity with recombinant mouse IL-10 | |
| Human | |
| 3586 | |
| <10% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| Absorbance at 450 nm minus absorbance (at 550 nm) | |
| 5 x 96 Tests | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 50 μL | |
| IL-10 | |
| 3 hrs |
| 15.36 to 600 pg/mL | |
| Human IL-10 ELISA Kit | |
| Colorimetric ELISA (TMB substrate) with Plates | |
| Plasma, Serum, Supernatant, Urine | |
| 5 x 96 microplate wells | |
| Colorimetric Microplate Reader | |
| CSIF,GVHDS,IL-10 A,TGIF,IL10 | |
| <10 | |
| HRP | |
| Intra-assay CV <10%; inter-assay CV <10% | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |