Learn More
Invitrogen™ Canine IL-10 ELISA Kit
ELISA
Supplier: Invitrogen™ ECIL10
Description
The Canine Interleukin 10 (Dog IL-10) ELISA quantitates Dog IL-10 in canine serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Dog IL-10. Principle of the method The Canine IL-10 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
IL-10 encodes a protein that acts as a cytokine and is primarily produced by monocytes, with some production by lymphocytes. This cytokine has various effects on immunoregulation and inflammation, including down-regulating the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. Additionally, IL-10 can block NF-kappa B activity and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice have suggested that this cytokine is an essential immunoregulator in the intestinal tract. Mutations in this gene have been associated with an increased susceptibility to HIV-1 infection and rheumatoid arthritis. Diseases associated with IL10 include Graft-Versus-Host Disease and Human Immunodeficiency Virus Type 1.Chemical Identifiers
| P48411 | |
| 0.1 ng/ml | |
| HRP | |
| Cytokines & Receptors | |
| This ELISA antibody pair detect canine IL-10. Other species not determined yet. | |
| ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
| 403628 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| Plasma 50 μL, Serum 50 μL, Supernatant 100 μL | |
| IL-10 | |
| 1 hrs 20 mins | |
| Solid-phase sandwich ELISA |
| 0.11 to 80 ng/mL | |
| 0.1 ng/ml to 80 ng/ml | |
| ELISA | |
| Plasma, Serum, Supernatant | |
| 96 assays | |
| Colorimetric Microplate Reader | |
| IL-10 | |
| <10 | |
| HRP | |
| Immunology | |
| 2°C to 8°C | |
| IL-10 | |
| 4 hrs 45 mins |
Specifications
| P48411 | |
| 0.1 ng/ml | |
| Solid-phase sandwich ELISA | |
| ELISA | |
| Plasma, Serum, Supernatant | |
| 96 assays | |
| ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
| 403628 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| 96 Tests | |
| Immunology | |
| 2°C to 8°C | |
| IL-10 | |
| 4 hrs 45 mins |
| 0.11 to 80 ng/mL | |
| 0.1 ng/ml to 80 ng/ml | |
| HRP | |
| Cytokines & Receptors | |
| This ELISA antibody pair detect canine IL-10. Other species not determined yet. | |
| Canine | |
| Colorimetric Microplate Reader | |
| IL-10 | |
| <10 | |
| HRP | |
| RUO | |
| Plasma 50 μL, Serum 50 μL, Supernatant 100 μL | |
| IL-10 | |
| 1 hrs 20 mins |