Learn More
PeproTech IL-1 beta Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: PeproTech 500P51BT50UG
Description
AA Sequence of recombinant protein: MVPIRQLHYR LRDEQQKSLV LSDPYELKAL HLNGQNINQQ VIFSMSFVQG EPSNDKIPVA LGLKGKNLYL SCVMKDGTPT LQLESVDPKQ YPKKKMEKRF VFNKIEVKSK VEFESAEFPN WYISTSQAEH KPVFLGNNSG QDIIDFTMES VSS. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-1beta. Anti-Murine IL-1beta-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect mIL-1beta by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine IL-1beta (500-P51) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mIL-1beta. Western Blot: To detect mIL-1beta by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mIL-1beta is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P51BT-1MG will be provided as 2 x 500 μg
Interleukin-1 beta (IL-1 beta) is a proinflammatory cytokine expressed by monocytes, macrophages, and dendritic cells. IL-1 beta is synthesized in response to inflammatory stimuli as a 31 kDa inactive pro-form that accumulates in the cytosol. Cleavage of pro-IL-1 beta into the active 17 kDa protein requires the activation of inflammasomes, which are multi-protein complexes that respond to pathogens, stress conditions, and other danger signals. Inflammasome activation triggers the processing of the caspase-1 precursor into its active form, which in turn cleaves pro-IL-1 beta. IL-1 beta lacks a signal sequence peptide for classical ER/Golgi pathway and is secreted alongside caspase-1 via an alternate and incompletely understood mechanism. Although IL-1 beta is most often secreted in its active form, secretion of the uncleaved protein may be detectable under some biological conditions. IL-1 beta signals through two receptors, IL-1RI and IL-1RII, both of which are shared with IL-1 alpha. IL-1 beta activity can be moderated by IL-1 Receptor Antagonist (IL-1RA), a protein produced by many cell types that blocks receptor binding through competitive inhibition. IL-1 beta play an important role in innate host defense by triggering the production of other proinflammatory cytokines in target cells and initiating acute-phase responses to infection and injury. Elevated levels of IL-1 beta have been associated with many chronic inflammatory conditions IL-1 beta neutralizing antibodies potential therapeutic value.Specifications
| IL-1 beta | |
| Polyclonal | |
| Biotin | |
| IL1B | |
| catabolin; cytokine; Hematopoietin 1 (H1); IFN beta inducing factor; il 1b; IL 1β; IL-1; IL1 B; IL-1 beta; IL-1 precursor; IL1B; IL-1B; IL1B1; IL1beta; IL-1beta; IL1-BETA; IL1F2; IL1β; ILN; Interleukin; interleukin 1 beta; Interleukin 1 beta precursor; interleukin 1, beta; interleukin 1, beta 1; interleukin 1-beta; interleukin*1*beta; Interleukin1 beta; interleukin-1 beta; interleukin-1 beta precursor; interleukin-1 beta precursor (AA -113 to 153); interleukin-1 beta proprotein; interleukin-1b; interleukin-1beta; interleukin-beta; LAF; lymphocyte proliferation-potentiating factor; Osteoclast activating factor (OAF); preinterleukin 1 beta; Pro interleukin 1 beta; prointerleukin-1 beta; pro-interleukin-1-beta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16176 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P10749 | |
| IL1B | |
| E.coli-derived Recombinant Murine IL-1beta. | |
| 50 μg | |
| Primary | |
| Mouse | |
| Antibody |