missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IKK epsilon Recombinant Rabbit Monoclonal Antibody (23GB2720)

Catalog No. MA552523
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552523 20 μL
MA552522 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552523 Supplier Invitrogen™ Supplier No. MA552523
Only null left

Rabbit Recombinant Monoclonal Antibody

IKK epsilon Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow

IKBKE is an activator of nuclear factor-kappa-B. Over expression of IKBKE has been observed in breast cancer cells, and suppression of IKBKE induces cell death. NF-kappa-B is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kappa-B mediates the expression of a great variety of genes in response to extracellular stimuli. NF-kappa-B is associated with Ikappa-B proteins in the cell cytoplasm, which inhibit NF-kappa-B activity. Ikappa-B is phosphorylated by Ikappa-B kinase (IKK) complex that contains IKK-a, IKK-b, and IKK-gamma. A novel molecule in the IKK complex was recently identified and designated IKK-iota/IKK-i. IKK epsilon is required for the activation of NF-kappa-B by PMA and T cell receptors but not by TNF-a and IL-1. IKK-iota/IKK-i message is expressed in a variety of tissues and is inducible by TNF-a, IL-1, and LPS.

Specifications

Antigen IKK epsilon
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB2720
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene IKBKE
Gene Accession No. Q14164, Q9R0T8
Gene Alias AW558201; I-kappa-B kinase epsilon; Ikbke; IKKE; IKK-E; IKKepsilon; IKK-epsilon; IKKI; IKK-i; IKK-related kinase epsilon; inducible I kappa-B kinase; inducible IkappaB kinase; inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon; inhibitor of kappaB kinase epsilon; inhibitor of nuclear factor kappa-B kinase subunit epsilon; KIAA0151; RP11-534L20.1
Gene Symbols IKBKE
Host Species Rabbit
Immunogen A synthesized peptide derived from human IKKi/IKKe (32-65AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363984, 56489, 9641
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less