missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IKK alpha Recombinant Rabbit Monoclonal Antibody (23GB1065)

Catalog No. MA554101
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA554101 20 μL
MA554100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA554101 Supplier Invitrogen™ Supplier No. MA554101
Only null left

Rabbit Recombinant Monoclonal Antibody

IKK alpha Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

CHUK (IKK alpha) is a serine/threonine kinase involved in signal transduction and cellular communication. It belongs to the cytokine activated protein complex that is an inhibitor of the NF-kappa B complex. IKK alpha along with IKK beta and a regulatory protein NEMO (IKK gamma) phosphorylates IkB proteins which targets the later for ubiquitin mediated degradation. It is involved with several cellular processes with extensive studies on its role in inflammation and cell survival.

Specifications

Antigen IKK alpha
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB1065
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CHUK
Gene Accession No. O15111, Q60680
Gene Alias AI256658; CHUK; Chuk1; component of inhibitor of nuclear factor kappa B kinase complex; conserved helix-loop-helix ubiquitous kinase; EC 2.7.11.10; Fbx24; Fbxo24; FLJ40509; I kappa B kinase 1; i kappa-B kinase alpha; ikappaB kinase; I-kappa-B kinase 1; I-kappa-B kinase 2; IkappaB kinase alpha; I-kappa-B kinase alpha; I-kappa-B kinase-alpha; IkB kinase alpha subunit; IkB kinase-alpha; ikBKA; IKBKB; IKK alpha; IKK1; IKK2; Ikka; IKK-A; IKK-a kinase; IKK-alpha; IKKB; IKK-B; IKK-beta; Inhibitor of nuclear factor kappa-B kinase subunit alpha; kinase beta; MGC131801; NFKBIKA; NFKBIKB; Nuclear factor NFkappaB inhibitor kinase alpha; Nuclear factor NF-kappa-B inhibitor kinase alpha; TCF16; TCF-16; Transcription factor 16
Gene Symbols CHUK
Host Species Rabbit
Immunogen A synthetic peptide of human IKK alpha (677-696AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1147, 12675
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less