missing translation for 'onlineSavingsMsg'
Learn More

IHC Select™ Secondary Goat Mouse IgG Goat anti-Mouse, Biotin, Polyclonal, Secondary Antibody, MilliporeSigma™

Catalog No. 21538MI
Change view
Click to view available options
Quantity:
15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21538MI 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21538MI Supplier MilliporeSigma Supplier No. 21538
Only null left

Goat Polyclonal Antibody

This Goat IgG antibody is validated for use in immunohistochemistry for the detection of the Mouse IgG protein.

Specifications

Antigen Goat anti-Mouse IgG
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Formulation Goat anti-Mouse IgG in PBS containing carrier protein, with 0.1% ProClin 300 as preservative.
Host Species Goat
Quantity 15 mL
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 2-8°C in undiluted format. Do not Freeze.
Form Liquid
Show More Show Less

For Research Use Only