missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ IHC Antigen Retrieval Solution - High pH (10X)

Catalog No. 5018783
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5018783 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5018783 Supplier Invitrogen™ Supplier No. 00495658

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for use during the heat induced epitope retrieval (HIER) step prior to immunohistochemistry on formalin-fixed paraffin embedded tissue sections. Use in combination with heat has been shown to restore the antigenicity of proteins modified during the formalin fixation of tissue.

  • Supplied as a 10x stock solution
  • Dilution of this buffer to 1x using reagent-grade water results in a 10 mM Tris, 1 mM EDTA solution, pH 9.0

Specifications

Product Type IHC Antigen Retrieval Solution
Quantity 500 mL
For Use With (Application) Immunohistochemistry (IHC)
Product Line eBioscience
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.