missing translation for 'onlineSavingsMsg'
Learn More

Iguratimod 98.0+%, TCI America™

Catalog No. p-7131669
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I094525MG 25 mg
I0945250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I094525MG Supplier TCI America Supplier No. I094525MG
Only null left

Chemical Identifiers

CAS 123663-49-0
Molecular Formula C17H14N2O6S
Molecular Weight (g/mol) 374.367
MDL Number MFCD00882374
InChI Key ANMATWQYLIFGOK-UHFFFAOYSA-N
Synonym N-(3-Formamido-4-oxo-6-phenoxy-4H-chromen-7-yl)methanesulfonamide, N-[7-(Methanesulfonamido)-4-oxo-6-phenoxy-4H-chromen-3-yl]formamide
PubChem CID 124246
IUPAC Name N-[7-(methanesulfonamido)-4-oxo-6-phenoxychromen-3-yl]formamide
SMILES CS(=O)(=O)NC1=C(C=C2C(=C1)OC=C(C2=O)NC=O)OC3=CC=CC=C3

Specifications

Melting Point 240°C
Color White-Yellow
Quantity 25 mg
Formula Weight 374.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Iguratimod