missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat IgM Monoclonal Antibody (HIS40), eBioscience™
GREENER_CHOICE

Catalog No. 5012773
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5012773 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5012773 Supplier Invitrogen™ Supplier No. 14099082
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Description: The HIS40 monoclonal antibody reacts with the μ heavy chain of rat IgM. It does not react with other classes of rat immunoglobulin including IgD, IgG or IgA. IgM is expressed intracellularly, during early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. Fluorochrome conjugated HIS40 is used as a detection secondary for rat IgM primary antibodies. Applications Reported: The HIS40 antibody has been reported for use in flow cytometric analysis, and immunohistochemical staining. Applications Tested: The HIS40 antibody has been tested by flow cytometric analysis of rat splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

Rat IgM is an immunoglobulin M isotype antibody produced by B cells in rats. It plays a crucial role in the immune system by recognizing and neutralizing antigens, such as bacteria and viruses. Rat IgM has been extensively studied and is widely used in research applications, including immunology, microbiology, and biotechnology. It is commonly used as a control in experiments, as well as a diagnostic tool for detecting infectious diseases and autoimmune disorders. Researchers have developed various techniques for producing and purifying Rat IgM, including hybridoma technology, protein A/G chromatography, and affinity chromatography. Rat IgM is a valuable tool in the field of immunology, and its use has contributed to a better understanding of the immune system and the development of new treatments for various diseases.

Specifications

Antigen Rat IgM
Applications Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone HIS40
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Igh-6
Gene Accession No. 0
Gene Alias Igh-6; IgM; immunoglobulin heavy chain 6; RGD1359202
Gene Symbols Igh-6
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299357
Target Species Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less