missing translation for 'onlineSavingsMsg'
Learn More

IgM, 5 Fc u-Specific PAb, anti-Human, Goat, Liq., MP Biomedicals

$292.05 - $772.12

Specifications

Antigen IgM, 5 Fc u-Specific
Classification Polyclonal
Conjugate Unconjugated
Host Species Goat
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN55097
View Documents
MP Biomedicals Inc
0855097
2 mL
Each for $292.05
Only null left
 
ICN55017
View Documents
MP Biomedicals Inc
0855017
5 mL
Each for $772.12
Only null left
 
Description

Description

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly

Specifications

Specifications

IgM, 5 Fc u-Specific
Unconjugated
Primary
Polyclonal
Goat
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'