missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ IgG, Species absorbed, Sheep anti-Mouse, HRP, Polyclonal,

Catalog No. AC111PMI
Change view
Click to view available options
Quantity:
500 μL
Conjugate:
HRP
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AC111PMI 500 μL HRP
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC111PMI Supplier MilliporeSigma™ Supplier No. AC111P

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sheep Polyclonal Antibody

Immunoglobulin G (IgG) is part of the family of glycoprotein immunoglobulins which stage immune responses against pathogenic organisms. IgG contributes to the immune response by mobilizing innate immune effector cells and inducing and pro-inflammatory responses. Such proinflammatory responses are mediated by Fc receptors which interact with sialylated Fc fragments on IgG molecules.

Specifications

Antigen IgG, Species absorbed
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate HRP
Formulation Purified sheep polyclonal antibody in PBS with StabilZyme™ HRP Conjugate Stabilizer and 0.05% Proclin.
Host Species Sheep
Immunogen Purified mouse IgG.
Purification Method Ammonium Sulfate
Quantity 500 μL
Regulatory Status RUO
Research Discipline Secondary & Control Antibodies
Primary or Secondary Secondary
Test Specificity Based on ELISA analysis using the unconjugated antibody (Mfr. No. AC111), this antibody reacts with whole molecule mouse IgG (IgG2>IgG2a>IgG3>>>IgG1) with minimal or no cross-reactivity toward mouse IgA or IgM.
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.