missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG, Polyclonal, MP Biomedicals
Specifications
Specifications
| Antigen | IgG |
| Classification | Polyclonal |
| Quantity | 2 mL |
| Primary or Secondary | Secondary |
| Antigen | IgG |
| Classification | Polyclonal |
| Quantity | 2 mL |
| Primary or Secondary | Secondary |