missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IGFBP-2 ELISA Kit

ELISA

$908.00

Specifications

Accession Number P47877
Assay Range 0.823 to 600 ng/mL
Assay Sensitivity 600 pg/mL
Calibration Range 0.6 ng/mL to 600 ng/mL
Assay Solid-phase sandwich ELISA
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
EMIGFBP2
Encompass
View Documents Promotion Details
Invitrogen™
EMIGFBP2
96 Tests
Pack for $908.00
Only null left
 
Description

Description

The Mouse IGF Binding Protein 2 (Ms IGFBP-2) ELISA quantitates Ms IGFBP-2 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IGFBP-2. Principle of the method The Mouse IGFBP-2 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IGFBP2 (insulin-like growth factor binding protein 2) is a member of the ISGBP family which bind various IGFs. IGFBP2 is overexpressed in a wide spectrum of cancers, including glioma, prostate cancer, synovial sarcoma, neuroblastoma, colon cancer, adrenocortical cancer, lung cancer, Wilms' tumor, and hepatoblastoma. The overexpression of IGFBP2 also correlates with the aggressiveness of some tumors. IGFBP2 activates the expression of matrix metalloprotease 2, which contributes to cell invasiveness.
Specifications

Specifications

P47877
600 pg/mL
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
Mouse
Colorimetric Microplate Reader
AI255832,IBP-2,Igfbp-2,mIGFBP-2
<10
HRP
Development
2°C to 8°C
IGFBP2
4 hrs 45 mins
0.823 to 600 ng/mL
0.6 ng/mL to 600 ng/mL
HRP
Growth Factors & Receptors
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CFS, IFN- gamma, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p40/p70, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP- 5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, PSelectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF RI, TNF RII, TPO, VCAM-1, VEGF.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
16008
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
AI255832, IBP-2, Igfbp-2, mIGFBP-2
1 hrs 20 mins
Videos
SDS
Documents

Documents