missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals IGF-1R Inhibitor, PPP

CAS: 477-47-4 | C22H22O8, C22H22O8 | 414.41 g/mol

Supplier:  Thermo Scientific Chemicals J65916MA

Catalog No. AAJ65916MA

Only null left
Explore available promotions

Description

Description

Picropodophyllotoxin is an insulin-like growth factor-I (IGF-I) receptor kinase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

10 mg
2°C to 8°C
477-47-4
414.40g/mol
Light Sensitive
Solid
COC1=CC(=CC(OC)=C1OC)[C@H]1[C@H]2[C@H](COC2=O)[C@@H](O)C2=CC3=C(OCO3)C=C12
414.41
White
Picropodophyllotoxin is an insulin-like growth factor-I (IGF-I) receptor kinase inhibitor
C22H22O8
Insulin-like Growth Factor-1 Receptor Inhibitor; Picropodophyllin
Signal Transduction Reagent
YJGVMLPVUAXIQN-HAEOHBJNSA-N
(10R,11S,15R,16R)-16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one

Chemical Identifiers

477-47-4
414.41
Insulin-like Growth Factor-1 Receptor Inhibitor; Picropodophyllin
COC1=CC(=CC(OC)=C1OC)[C@H]1[C@H]2[C@H](COC2=O)[C@@H](O)C2=CC3=C(OCO3)C=C12
C22H22O8
YJGVMLPVUAXIQN-HAEOHBJNSA-N
(10R,11S,15R,16R)-16-hydroxy-10-(3,4,5-trimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only