Learn More
IGEPAL™ CA-630 , MilliporeSigma™ Supelco™
IGEPAL™ CA-630 is a nonionic, non-denaturing detergent suitable for solubilization, isolation and purification of membrane protein complexes. It is chemically indistinguishable from Nonidet P-40.
Supplier: Merck Emd Millipore 56741250MLF
Specifications
| 9002-93-1 | |
| 1.06 g/mL (at 20°C) | |
| 250 mL |
| (C2H4O)nC14H22O | |
| Nonidet(TM) P 40; Nonylphenyl-polyethylenglycol; Octylphenoxy poly(ethyleneoxy)ethanol, branched; Octylphenoxy poly(ethyleneoxy)ethanol, branched | |
| n20/D 1.492 |
IGEPAL is a registered trademark of Rhodia Operations