missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IGEPAL™ CA-630 , MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 9002-93-1 |
| Linear Formula | (C2H4O)nC14H22O |
| Density | 1.06 g/mL (at 20°C) |
| Synonym | Nonidet(TM) P 40; Nonylphenyl-polyethylenglycol; Octylphenoxy poly(ethyleneoxy)ethanol, branched; Octylphenoxy poly(ethyleneoxy)ethanol, branched |
| Quantity | 50 mL |
| Refractive Index | n20/D 1.492 |
IGEPAL is a registered trademark of Rhodia Operations