Learn More
Invitrogen™ Mouse IgE Uncoated ELISA Kit with Plates
ELISA
$440.00 - $1,574.00
Specifications
| Assay Range | 4 to 250 ng/mL |
|---|---|
| Assay Sensitivity | 4 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
501125118
|
Invitrogen™
885046076 |
20 x 96 Tests |
Each for $1,574.00
|
|
|||||
|
501125120
|
Invitrogen™
885046086 |
10 x 96 Tests |
Each for $1,140.00
|
|
|||||
|
501128838
|
Invitrogen™
885046022 |
2 x 96 Tests |
Each for $440.00
|
|
|||||
Description
The Mouse Immunoglobulin E (IgE) Uncoated ELISA Kit contains pre-matched antibody pairs ,plates, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IgE. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
IgE is one of the five classes of immunoglobulins found in mammals. Monomers consist of two heavy (epsilon) chains with 4 Ig-like constant domains, and two light chains. IgE plays an essential role in type I hypersensitivity (allergic responses such as hay fever, asthma, hives, anaphylatic shock) by binding to Fc receptors on basophils and mast cells. Crosslinking of IgE bound to Fc receptors induces degranulation of mast cells and basophils that promote allergic manifestations. IgE is also functionally important for immunity against parasites.Specifications
| 4 to 250 ng/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| ELISA | |
| 380792 | |
| HRP | |
| Plasma 10 μL, Serum 10 μL, Supernatant 50 μL | |
| Immunoglobulin E | |
| 1 hrs 45 mins |
| 4 ng/mL | |
| ELISA | |
| Mouse | |
| Colorimetric Microplate Reader | |
| Capture Antibody: Pre-titrated, Purified antibody, Detection Antibody: Pre-titrated, HRP-conjugated antibody, Standard: Recombinant protein for generating standard curve and calibrating samples, 10X Coating Buffer: Buffer for plating the Capture Antibody, 5X ELISA/ELISPOT Diluent: Buffer for blocking and diluting the Detection Antibody and Enzyme, Substrate: 1X TMB Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: High affinity 96-well plate (included with product Cat. Nos. ending in suffixes -22, -76, -86) | |
| RUO | |
| 2°C to 8°C | |
| Mouse IgE | |
| 24 hrs 30 mins |