Learn More
Invitrogen™ IgE Monoclonal Antibody (23G3), Biotin, eBioscience™, Invitrogen™

Description
Description: The 23G3 monoclonal antibody reacts with the epsilon heavy chain of mouse IgE. It does not react with other classes of mouse immunoglobulin including IgD, IgG, IgA, or IgM. Applications Reported: The 23G3 antibody has been reported for use in flow cytometric analysis, and ELISA. Applications Tested: The 23G3 antibody has been tested by ELISA and can be used at 1:5000-1:10000. This 23G3 antibody has also been tested by flow cytometric analysis of IGEL cells, and can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
Specifications
Specifications
| Antigen | Mouse IgE |
| Applications | ELISA, Flow Cytometry |
| Classification | Monoclonal |
| Clone | 23G3 |
| Concentration | 0.5 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Gene | Ighe |
| Gene Accession No. | P06336 |
| Gene Alias | Gm900; Ighe; Immunoglobulin E; Immunoglobulin heavy constant epsilon; ImmunoglobulinE |
| Show More |
For Research Use Only.