missing translation for 'onlineSavingsMsg'
Learn More

Ifosfamide

Catalog No. AC461350050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461350050 5 g
AC461350010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461350050 Supplier Thermo Scientific Chemicals Supplier No. 461350050

missing translation for 'validMainframeMsg'

CAS: 3778-73-2 | C7H15Cl2N2O2P | 261.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3778-73-2
Molecular Formula C7H15Cl2N2O2P
Molecular Weight (g/mol) 261.09
InChI Key HOMGKSMUEGBAAB-UHFFFAOYSA-N
Synonym ifosfamide, isophosphamide, iphosphamide, isofosfamide, ifosfamid, mitoxana, naxamide, iphosphamid, isoendoxan, ifex
PubChem CID 3690
ChEBI CHEBI:5864
IUPAC Name N,3-bis(2-chloroethyl)-2-oxo-1,3,2$l^{5}-oxazaphosphinan-2-amine
SMILES C1CN(P(=O)(OC1)NCCCl)CCCl

Specifications

Melting Point 39.0°C to 42.0°C
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 261.09
Percent Purity ≥97.5% (ELSD) (HPLC)
Chemical Name or Material Ifosfamide
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/
EINECSNumber 223-237-3

RUO – Research Use Only