missing translation for 'onlineSavingsMsg'
Learn More

IFNW1, Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89020236
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89020236 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89020236 Supplier Abnova Corporation Supplier No. H00003467B01P
Only null left

Mouse Polyclonal Antibody

Sequence: MALLFPLLAALVMTSYSPVGSLGCDLPQNHGLLSRNTLVLLHQMRRISPFLCLKDRRDFRFPQEMVKGSQLQKAHVMSVLHEMLQQIFSLFHTERSSAAWNMTLLDQLHTGLHQQLQHLETCLLQVVGEGESAGAISSPALTLRRYFQGIRVYLKEKKYSDCAWEVVRMEIMKSLFLSTNMQERLRSKDRDLGSS

Specifications

Antigen IFNW1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene interferon, omega 1
Gene Accession No. NM_002177
Gene Alias -
Gene Symbols IFNW1
Host Species Mouse
Immunogen IFNW1 (NP_002168.1, 1 a.a. ∼ 195 a.a) full-length human protein.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3467
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Show More Show Less