missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFN omega Monoclonal Antibody (K16289_11E9), Invitrogen™

Catalog No. MA556209
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556209 200 μL
MA555829 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556209 Supplier Invitrogen™ Supplier No. MA556209
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: GLLSRNTLVL LHQMRRISPF LCLKDRRDFR FPQEMVKGSQ LQKAHVMSVL HEMLQQIFSL FHTERSSAAW NMTLLDQLHT GLHQQLQHLE TCLLQVVGEG ESAGAISSPA LTLRRYFQGI RVYLKEKKYS DCAWEVVRME IMKSLFLSTN MQERLRSKDR

IFN-omega is a type I interferon that can be induced by virus-infected leukocytes. Members of the type I interferon family, which includes IFN-alpha, IFN-beta, and IFN-omega, signal through the IFNAR-1/IFNAR-2 receptor complex, and exert antiviral and antiproliferative activities. IFN-omega exhibits about 75% sequence homology with IFN-alpha, and contains two conserved disulfide bonds that are necessary for full biological activity.

Specifications

Antigen IFN omega
Applications Western Blot
Classification Monoclonal
Clone K16289_11E9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene IFNW1
Gene Accession No. P05000
Gene Alias IFN o; IFN ω; IFNo; IFNomega; IFN-omega 1, interferon omega-1; IFNW; IFNW1; IFNω; Interferon; Interferon alpha-II-1; Interferon o; Interferon OMEGA; interferon omega 1; interferon omega-1; Interferon ω; interferon, omega 1
Gene Symbols IFNW1
Host Species Mouse
Immunogen Recombinant human IFNW1 (immunogen range is 31-190).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3467
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less