missing translation for 'onlineSavingsMsg'
Learn More

ProSci IFN omega 1 Recombinant Protein

Catalog No. 16593904
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16593904 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16593904 Supplier ProSci Supplier No. 918640050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recombinant Protein

This recombinant protein can be used for biological assays. For research use only.

Interferon omega-1 is also known as Interferon alpha-II-1and IFNW1. It is a Secreted protein that in humans is encoded by the IFNW1 gene. IFNW1 belongs to the alpha/beta interferon family. Type I IFNs consist of IFN alpha, beta , τ, and ω and bind to the type I IFN receptor, whereas IFN- gamma is the only type II IFN and is specific for the type II IFN receptor. IFNW1 is a recently discovered protein structurally related to IFN-alpha and -beta. It has been shown that IFN-omega 1 similar to that of other human class I IFNs; potent antiviral activity was also observed on cells of bovine and ovine but not of equine or murine origin.

Specifications

Accession Number P05000
For Use With (Application) Not Available
Gene ID (Entrez) 3467
Molecular Weight (g/mol) 21.1 kDa
Quantity 0.05 mg
Source Human Cells
Common Name interferon, omega 1
Gene Symbol IFNW1
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag C-6 His tag
Content And Storage Lyophilized protein should be stored at -20°C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4°C to 7°C for 2-7 days. Aliquots of reconstituted samples are stable (at -20°C for 3 months.)
Form Lyophilized
Purity or Quality Grade >95% as determined by reducing SDS-PAGE. Endotoxin level less than 0.1 ng/μg (1 IEU/μg) as determined by LAL test.
Synonym Interferon omega-1 is also known as Interferon alpha-II-1 and IFNW1., IFNW1
Show More Show Less