missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IFN epsilon Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149855
Description
IFN epsilon Polyclonal Antibody for IHC (P), Western Blot
Type I interferon required for maintaining basal levels of IFN-regulated genes, including 2'-5'-oligoadenylate synthetase, IRF7 and ISG15, in the female reproductive tract. Directly mediates protection against viral and bacterial genital infections. [UniProt].Specifications
| IFN epsilon | |
| Polyclonal | |
| Unconjugated | |
| IFNE | |
| IFNE; IFN-E; Ifne1; IFN-epsilon; Ifnt1; Ifn-tau-1; Infe1; Interferon; Interferon epsilon; interferon epsilon 1; interferon epsilon-1; Interferon tau-1; interferon, epsilon; interferon-epsilon; PRO655; UNQ360/PRO655 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 338376 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q86WN2 | |
| IFNE | |
| Recombinant Interferon Epsilon (IFNe) (Gln30-Arg208). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |