missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFN beta Recombinant Rabbit Monoclonal Antibody (23GB6230)

Catalog No. MA553003
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553003 20 μL
MA553002 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553003 Supplier Invitrogen™ Supplier No. MA553003
Only null left

Rabbit Recombinant Monoclonal Antibody

IFN beta Recombinant Monoclonal Antibody for ICC/IF, Western Blot

Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.

Specifications

Antigen IFN beta
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6230
Concentration 0.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ifnb1
Gene Accession No. P01574, P01575
Gene Alias Fibroblast interferon; Ifb; IFF; IFN b; IFN β; IFNB; Ifnb1; IFNbeta; IFN-beta; IFNβ; Interferon; Interferon b; Interferon beta; interferon beta 1; interferon beta 1, fibroblast; interferon beta protein; interferon, beta 1; interferon, beta 1, fibroblast; interferon-beta; RP11-113D19.1
Gene Symbols Ifnb1
Host Species Rabbit
Immunogen A synthesized peptide derived from human Interferon beta (50-110AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15977, 3456
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less