missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFN beta Polyclonal Antibody

Catalog No. PI314101
Change view
Click to view available options
Quantity:
2x10^4 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI314101 2x10^4 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI314101 Supplier Invitrogen™ Supplier No. LSG314101
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Specificity: Neutralizes human interferon beta. Activity: 2.8x10^5 NU/mL. Interferon was titrated with the use of the cytopathic effect inhibition assay using A549 cells with EMCV, where 1 unit/mL of interferon was necessary to produce an endpoint of 50%. One neutralization unit is defined as the amount of serum required to neutralize one unit of human IFN beta to an endpoint of 50%. Store at -20°C for short-term use (less than 6 months). For long-term storage, aliquot and store at -70°C or below for retention of full activity. Refreezing should be done on dry ice or in a dry ice/alcohol bath. Further dilution of the product should be in buffers containing protein such as 0.1% BSA. Packaged by and distributed for PBL Assay Science.

Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.

Specifications

Antigen IFN beta
Applications Neutralization
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Whole serum, PBS with no preservative
Gene Ifnb1
Gene Accession No. P01574
Gene Alias Fibroblast interferon; Ifb; IFF; IFN b; IFN β; IFNB; Ifnb1; IFNbeta; IFN-beta; IFNβ; Interferon; Interferon b; Interferon beta; interferon beta 1; interferon beta 1, fibroblast; interferon beta protein; interferon, beta 1; interferon, beta 1, fibroblast; interferon-beta; RP11-113D19.1
Gene Symbols Ifnb1
Host Species Rabbit
Immunogen Recombinant human interferon beta.
Quantity 2x10^4 units
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3456
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less