missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IFN beta Monoclonal Antibody (A1 (IFNb)), FITC, eBioscience™

Mouse Monoclonal Antibody

Supplier:  Invitrogen™ BMS1044FI

Catalog No. 501125239


Only null left
Explore available promotions

Description

Description

Description: The IFNb/A1 clone recognizes natural human IFN-beta and recombinants beta1a and beta1b. It is capable of neutralizing IFN-beta. Interferon beta is synthesized and secreted by fibroblasts and many other cell types in response to pathogens. IFN-beta binding to type I interferon receptors induces the upregulation of IRF-7 and activation of Rnase L. IRF-7 can exert a positive feedback on IFN-beta production. RNase L cleaves both viral and cellular single stranded mRNA, thereby limiting viral replication and dissemination. Applications Tested: ELISA, Neutralization tests, Western Blotting. Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.

Type I Interferons (IFN-alpha/beta) are produced primarily in response to viral infection by Natural IFN-producing cells (IPCs) as part of the host immune response. IFNs can also inhibit the development of tumors. IFN-beta binding results in the activation of the tyrosine kinases Jak1 and Tyk2, phosphorylation of members of the STAT family of transcription factors, and the transcription and expression of the immune response genes. More recently, several members of the toll-like receptor (TLR) family were found to stimulate the production IFN-beta. IFN-beta is currently used clinically for treatment of tumors, infections and multiple sclerosis.
Specifications

Specifications

IFN beta
Monoclonal
0.1 mg/mL
TBS with 1% BSA and 0.02% sodium azide; pH 7.2-7.4
P01574
Ifnb1
Affinity chromatography
RUO
3456
4°C, store in dark
Liquid
ELISA, Functional Assay, Western Blot
A1 (IFNb)
FITC
Ifnb1
Fibroblast interferon; Ifb; IFF; IFN b; IFN β; IFNB; Ifnb1; IFNbeta; IFN-beta; IFNβ; Interferon; Interferon b; Interferon beta; interferon beta 1; interferon beta 1, fibroblast; interferon beta protein; interferon, beta 1; interferon, beta 1, fibroblast; interferon-beta; RP11-113D19.1
Mouse
100 μg
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions