missing translation for 'onlineSavingsMsg'
Learn More

Ifenprodil hemitartrate, 99%

Catalog No. AAJ60932MF
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60932MF 50 mg
AAJ60932MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60932MF Supplier Thermo Scientific Chemicals Supplier No. J60932MF

NMDA receptor antagonist, acting at the polyamine site | CAS: 23210-56-2 | C21H27NO2 | 325.452 g/mol

NMDA receptor antagonist, acting at the polyamine site

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23210-56-2
Molecular Formula C21H27NO2
Molecular Weight (g/mol) 325.452
MDL Number MFCD01529999
InChI Key UYNVMODNBIQBMV-UHFFFAOYSA-N
Synonym ifenprodil, vadilex, ifenprodil tartrate, dilvax, creocral, ifenprodil inn:dcf, ifenprodilum inn-latin, unii-r8oe3p6o5s, 4-2-4-benzylpiperidin-1-yl-1-hydroxypropyl phenol, r8oe3p6o5s
PubChem CID 3689
IUPAC Name 4-[2-(4-benzylpiperidin-1-yl)-1-hydroxypropyl]phenol
SMILES CC(C(C1=CC=C(C=C1)O)O)N2CCC(CC2)CC3=CC=CC=C3

Specifications

Color White
Quantity 50 mg
Merck Index 14,4895
Formula Weight 400.49
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Ifenprodil hemitartrate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 245-491-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only