accessibility menu, dialog, popup

UserName

H3COHOHN

CAS RN 23210-56-2

IUPAC Name: 4-[2-(4-benzylpiperidin-1-yl)-1-hydroxypropyl]phenol
Synonyms: ifenprodil 艾芬地尔
Molecular Weight (g/mol): 325.45
Molecular Formula: C21H27NO2
InChi Key: UYNVMODNBIQBMV-UHFFFAOYNA-N
SMILES: CC(C(O)C1=CC=C(O)C=C1)N1CCC(CC2=CC=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results