missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IDNK Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566169
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566169 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566169 Supplier Invitrogen™ Supplier No. PA566169
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: RDILTQGKDGV ALKCEESGKE AKQAEMQLLV VHLSGSFEVI SGRLLKREGH Highest antigen sequence identity to the following orthologs - mouse 53%, rat 55%.

The function of this protein remains unknown.

Specifications

Antigen IDNK
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Idnk
Gene Accession No. Q5T6J7
Gene Alias 5133401N09Rik; bA522I20.2; BCRG2; C9orf103; glucokinase-like protein; gluconate kinase; gluconokinase-like protein; hGntK; Idnk; idnK gluconokinase homolog (E. coli); IDNK, gluconokinase; idnK, gluconokinase homolog; KIAA1824; probable gluconokinase; WDR22
Gene Symbols Idnk
Host Species Rabbit
Immunogen Recombinant Human IDNK. Recombinant protein control fragment (Product #RP-106855).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 414328
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less