missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ID4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595723
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595723 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595723 Supplier Invitrogen™ Supplier No. PA595723
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: MKAVSPVRPS GRKAPSGCGG GELALRCLAE HGHSLGGSAA AAAAAAAARC KAAEAAADEP ALCLQCDMND CYSRLRRLVP TIPPNKKVSK VEILQHVIDY.

ID4 is transcription factors containing a basic helix-loop-helix (bHLH) motif regulate expression of tissue-specific genes in a number of mammalian and insect systems. DNA-binding activity of the bHLH proteins is dependent on formation of homo- and/or heterodimers. Dominant-negative HLH proteins encoded by Id-related genes, such as ID4, also contain the HLH-dimerization domain but lack the DNA-binding basic domain. Consequently, Id proteins inhibit binding to DNA and transcriptional transactivation by heterodimerization with bHLH proteins.

Specifications

Antigen ID4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Id4
Gene Accession No. P41139
Gene Alias bHLHb27; cb547; Class B basic helix-loop-helix protein 27; DNA-binding protein inhibitor ID-4; fb77d04; hypothetical protein LOC664761; ID4; Id-4; Idb4; Inhibitor of differentiation 4; inhibitor of DNA binding 4; inhibitor of DNA binding 4, dominant negative helix-loop-helix protein; inhibitor of DNA binding 4, HLH protein; sb:cb547; wu:fb77d04; zgc:123214
Gene Symbols Id4
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ID4 (NP_0015371).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15904, 291023
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less