missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ID2 Monoclonal Antibody (ILCID2), PE-Cyanine7, eBioscience™, Invitrogen™

Catalog No. 25947580
Encompass
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25947580 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 25947580 Supplier Invitrogen™ Supplier No. 25947580
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Mouse Monoclonal Antibody

Description: This ILCID2 monoclonal antibody recognizes mouse and human ID2. This ILCID2 antibody requires Foxp3/Transcription Factor Staining Buffer set and will perform poorly when used with standard IC Fixation & Permeabilization Buffer Set and protocol. Applications Reported: This ILCID2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This ILCID2 antibody has been tested by flow cytometric analysis of mouse splenocytes using the Foxp3/Transcription Factor Staining Buffer Set (Product No. 00-5523) and protocol. Please refer to Best Protocols: Protocol B: One step protocol for (nuclear) intracellular proteins located under the Resources Tab online. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product No. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product No.

The inhibitor of DNA binding 2 (ID2) protein is essential for promoting the development of a number of cell types including natural killer cell, erythroid precursors, as well as, CD8+ and CD4+ T-cell. A lack of ID2 promotes B-cell development. The chief role of this protein is to associate with ubiquitously-expressed E proteins, and other proteins of the basic helix-loop-helix (HLH) family, preventing them from binding to DNA and activating transcription. Activation of ID2 is critical for the development of the natural killer cell lineage.

Specifications

Antigen ID2
Applications Flow Cytometry
Classification Monoclonal
Clone ILCID2
Concentration 0.2 mg/mL
Conjugate PE-Cyanine7
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene ID2
Gene Accession No. P41136, Q02363
Gene Alias Ac2-300; AI255428; bHLHb26; C78922; cell growth-inhibiting gene 8; class B basic helix-loop-helix protein 26; DNA-binding protein inhibitor ID2; DNA-binding protein inhibitor ID-2; GIG8; helix-loop-helix protein ID2; Id2; Id-2; ID2A; ID2H; Idb2; Inhibitor of differentiation 2; inhibitor of DNA binding 2; inhibitor of DNA binding 2, dominant negative helix-loop-helix protein; inhibitor of DNA binding 2, HLH protein; MGC26389; OTTHUMP00000140258; OTTHUMP00000200297
Gene Symbols ID2
Host Species Mouse
Immunogen Purified recombinant fragment of human ID2, corresponding to amino acids 1-134 expressed in E. coli.
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15902, 3398
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less